Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-amino-4-methylthiophene-3-carboxylate
021476-5G
021476-25G
FLUO021476-5GEA
96.5
EUR
FLUO021476-5G
FLUO021476-25G
Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Formel:
C₈H₁₁NO₂S Molecular Weight: 185.25 g/mol |
MDL:
MFCD00051669 CAS-Nummer: 43088-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...