Ethyl-2-amino-4,5-dimethyl-3-thiophencarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate

017433-10G 017433-25G 017433-50G
FLUO017433-10GEA 101 EUR
FLUO017433-10G FLUO017433-25G FLUO017433-50G
Ethyl-2-amino-4,5-dimethyl-3-thiophencarboxylat
Ethyl-2-amino-4,5-dimethyl-3-thiophencarboxylat
Formel: C₉H₁₃NO₂S
Molecular Weight: 199,27 g/mol
MDL: MFCD00085051
CAS-Nummer: 4815-24-1
EINECS: 225-387-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR