5-Chlor-3-methylbenzo[b]thiophen
Lieferant: FLUOROCHEM
005326-1G
005326-5G
005326-25G
FLUO005326-1GEA
31.1
EUR
FLUO005326-1G
FLUO005326-5G
FLUO005326-25G
5-Chlor-3-methylbenzo[b]thiophen
5-Chlor-3-methylbenzo[b]thiophen
Formel:
C₉H₇ClS Molecular Weight: 182,67 g/mol |
MDL:
MFCD00052506 CAS-Nummer: 19404-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...