(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

M-Clarity™ Qualitätsstufe = MQ200

332593-100G 332593-25G 332593-5G
SIAL332593-100GEA 393 EUR
SIAL332593-100G SIAL332593-25G SIAL332593-5G
(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®
(2-Hydroxylpropyl)-β-cyclodextrin
Formel: C₆₃H₁₁₂O₄₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16621721
CAS-Nummer: 128446-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR