(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

M-Clarity™ Qualitätsstufe = MQ200

332607-100G 332607-25G 332607-500G 332607-5G
SIAL332607-100GEA 337 EUR
SIAL332607-100G SIAL332607-25G SIAL332607-500G SIAL332607-5G
(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®
(2-Hydroxylpropyl)-β-cyclodextrin
Formel: C₆₃H₁₁₂O₄₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16621721
CAS-Nummer: 128446-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR