(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
M-Clarity™ Qualitätsstufe = MQ200
C0926-10G
C0926-5G
SIALC0926-10GEA
1120
EUR
SIALC0926-10G
SIALC0926-5G
(2-Hydroxylpropyl)-β-cyclodextrin, Sigma-Aldrich®
(2-Hydroxylpropyl)-β-cyclodextrin
Formel:
C₆₃H₁₁₂O₄₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16621721 CAS-Nummer: 128446-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...