(2-Hydroxylpropyl)-γ-cyclodextrin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-Hydroxypropyl)-γ-Cyclodextrin, HGC
M-Clarity™ Qualitätsstufe = MQ100
390704-25G
390704-5G
SIAL390704-25GEA
164
EUR
SIAL390704-25G
SIAL390704-5G
(2-Hydroxylpropyl)-γ-cyclodextrin, Sigma-Aldrich®
(2-Hydroxylpropyl)-γ-cyclodextrin
Formel:
C₇₂H₁₂₈O₄₈ Molecular Weight: 1761,7600 g/mol |
MDL:
MFCD00148890 CAS-Nummer: 128446-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...