2,6-Di-O-methyl-β-cyclodextrin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, Heptakis(2,6-di-o-methyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin
M-Clarity™ Qualitätsstufe = MQ200
H0513-1G
H0513-5G
SIALH0513-1GEA
35.8
EUR
SIALH0513-1G
SIALH0513-5G
2,6-Di-O-methyl-β-cyclodextrin, Sigma-Aldrich®
2,6-Di-O-methyl-β-cyclodextrin
Formel:
C₅₆H₉₈O₃₅ Molecular Weight: 1331,3563 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011616 CAS-Nummer: 51166-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...