γ-Cyclodextrin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
M-Clarity™ Qualitätsstufe = MQ200
C4892-1G
C4892-5G
SIALC4892-1GEA
296
EUR
SIALC4892-1G
SIALC4892-5G
γ-Cyclodextrin, Sigma-Aldrich®
γ-Cyclodextrin
Formel:
C₄₈H₈₀O₄₀ Molecular Weight: 1297.1248 g/mol |
MDL:
MFCD00009595 CAS-Nummer: 17465-86-0 EINECS: 241-482-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...