γ-Cyclodextrin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin

M-Clarity™ Qualitätsstufe = MQ200

C4930-100MG C4930-1G
SIALC4930-100MGEA 240 EUR
SIALC4930-100MG SIALC4930-1G
γ-Cyclodextrin, Sigma-Aldrich®
γ-Cyclodextrin
Formel: C₄₈H₈₀O₄₀
Molecular Weight: 1297.1248 g/mol
MDL: MFCD00009595
CAS-Nummer: 17465-86-0
EINECS: 241-482-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR