β-Cyclodextrin Hydrat

Lieferant: Thermo Fisher Scientific

A14529.06 A14529.14 A14529.22
A14529.06EA 27.6 EUR
A14529.06 A14529.14 A14529.22
β-Cyclodextrin Hydrat
β-Cyclodextrin Hydrat

CAS No.: 68168-23-0 Molecular Formula: C42H70O35.xH2O Formula Weight: 1134.98(anhy) Melting Point: 288° dec. Beilstein Reference: 5915513 Merck Reference: 14,2718 Optical Rotation: +142° (c=1.5 in water) MDL No.: MFCD00150811

Formel: C₄₂H₇₀O₃₅·H₂O
Molecular Weight: 1134.98 g/mol (anhy)
Schmelzpunkt: 288 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150811
CAS-Nummer: 68168-23-0
EINECS: 231-493-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR