(±)-Indolin-2-carbonsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Indoline-2-carboxylic acid
B25450.06
B25450.14
B25450.06EA
57.7
EUR
B25450.06
B25450.14
(±)-Indolin-2-carbonsäure 95%
(±)-Indolin-2-carbonsäure
Purity: 95%
CAS No.: 78348-24-0
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: ca 168° dec.
MDL No.: MFCD00010635
Formel:
C₉H₉NO₂ Molecular Weight: 163,18 g/mol Schmelzpunkt: ∼ 168 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010635 CAS-Nummer: 78348-24-0 EINECS: 278-899-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...