(±)-Indolin-2-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: Indoline-2-carboxylic acid

CA-4489-5G CA-4489-25G CA-4489-100G
COBBCA-4489-5GEA 38.5 EUR
COBBCA-4489-5G COBBCA-4489-25G COBBCA-4489-100G
(±)-Indolin-2-carbonsäure
(±)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00010635
CAS-Nummer: 78348-24-0
EINECS: 278-899-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR