DL-Menthol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
A18098.0B
A18098.18
A18098.22
A18098.30
A18098.0BEA
209
EUR
A18098.0B
A18098.18
A18098.22
A18098.30
DL-Menthol ≥98%
DL-Menthol
Purity: 98+%
CAS No.: 89-78-1
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 93°(199°F)
Refractive Index: 1.4615
Odour: Sweet
Melting Point: 33-36°
Boiling Point: 212-216°
Beilstein Reference: 3194263
Merck Reference: 14,5837
MDL No.: MFCD00001484
Formel:
C₁₀H₂₀O Molecular Weight: 156,27 g/mol Siedepunkt: 212…216 °C Schmelzpunkt: 33…36 °C Dichte: 0,890 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001484 CAS-Nummer: 89-78-1 EINECS: 201-939-0 Merck Index: 14,5837 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...