DL-Menthol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol

M-Clarity™ Qualitätsstufe = MQ200

M2772-5G-A M2772-100G-A M2772-500G-A
SIALM2772-5G-AEA 27.7 EUR
SIALM2772-5G-A SIALM2772-100G-A SIALM2772-500G-A
DL-Menthol, Sigma-Aldrich®
DL-Menthol
Formel: C₁₀H₂₀O
Molecular Weight: 156,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001484
CAS-Nummer: 89-78-1
EINECS: 201-939-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR