Vitamin K1 (Phytomenadion, Phyllochinon), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione

95271-250MG 95271-1G
SIAL95271-250MGEA 27.8 EUR
SIAL95271-250MG SIAL95271-1G
Vitamin K1 (Phytomenadion, Phyllochinon), Sigma-Aldrich®
Vitamin K1 (Phytomenadion, Phyllochinon)
Formel: C₃₁H₄₆O₂
Molecular Weight: 450,6958 g/mol
MDL: MFCD00214063
CAS-Nummer: 84-80-0
EINECS: 201-564-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR