5-Fluororotsäure Monohydrat

Lieferant: FLUOROCHEM

Synonyme: 5-FOA

003234-1G 003234-10G 003234-50G 003234-25G 003234-5G
FLUO003234-1GEA 48.8 EUR
FLUO003234-1G FLUO003234-10G FLUO003234-50G FLUO003234-25G FLUO003234-5G
5-Fluororotsäure Monohydrat
5-Fluororotsäure Monohydrat
Formel: C₅H₃FN₂O₄·1H₂O
Molecular Weight: 174,0 g/mol (anhy)
MDL: MFCD00150658
CAS-Nummer: 220141-70-8
EINECS: 211-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR