5-Fluororotsäure Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid, 5-Fluorouracil-4-carboxylic acid

F5013-250MG F5013-50MG F5013-1G
SIALF5013-250MGEA 620 EUR
SIALF5013-250MG SIALF5013-50MG SIALF5013-1G
5-Fluororotsäure Hydrat, Sigma-Aldrich®
5-Fluororotsäure Hydrat
Formel: C₅H₃FN₂O₄·H₂O
Molecular Weight: 174,04 g/mol (anhy)
MDL: MFCD00150658
CAS-Nummer: 207291-81-4
EINECS: 211-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR