2-Thienylacetylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

L09323.06 L09323.14
L09323.06EA 27.4 EUR
L09323.06 L09323.14
2-Thienylacetylchlorid ≥98%
2-Thienylacetylchlorid

Purity: 98% CAS No.: 39098-97-0 Molecular Formula: C6H5ClOS Formula Weight: 160.62 Flash Point: 101°(213°F) Refractive Index: 1.5513 Boiling Point: 131-132°/90mm Beilstein Reference: 114547 UN No.: 3265 MDL No.: MFCD00005456

Formel: C₆H₅ClOS
Molecular Weight: 160,62 g/mol
Siedepunkt: 131…132 °C (90 mmHg)
Dichte: 1,303
Flammpunkt: 101 °C (213 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00005456
CAS-Nummer: 39098-97-0
EINECS: 254-290-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR