2-Thienylacetylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

003050-25G 003050-100G
FLUO003050-25GEA 79.1 EUR
FLUO003050-25G FLUO003050-100G
2-Thienylacetylchlorid
2-Thienylacetylchlorid
Formel: C₆H₅ClOS
Molecular Weight: 160.62 g/mol
MDL: MFCD00005456
CAS-Nummer: 39098-97-0
EINECS: 254-290-0
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR