4-(2-Thienyl)buttersäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Thiophen-2-yl)butanoic acid

L07563.03 L07563.06
L07563.03EA 26.6 EUR
L07563.03 L07563.06
4-(2-Thienyl)buttersäure ≥98%
4-(2-Thienyl)buttersäure

Purity: 98% CAS No.: 4653-11-6 Molecular Formula: C8H10O2S Formula Weight: 170.23 Flash Point: 110°(230°F) Refractive Index: 1.5320 Boiling Point: 122°/0.3mm Beilstein Reference: 127982 MDL No.: MFCD00005463

Formel: C₈H₁₀O₂S
Molecular Weight: 170,23 g/mol
Siedepunkt: 122 °C (0,3 mmHg)
Dichte: 1,169
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005463
CAS-Nummer: 4653-11-6
EINECS: 225-090-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR