2-Methoxy-6-(tributylstannyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
721115-1G
721115-5G
SIAL721115-1GEA
126
EUR
SIAL721115-1G
SIAL721115-5G
2-Methoxy-6-(tributylstannyl)pyridin, Sigma-Aldrich®
2-Methoxy-6-(tributylstannyl)pyridin
Formel:
C₁₈H₃₃NOSn |
MDL:
MFCD09025803 CAS-Nummer: 164014-94-2 UN: 2788 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...