2-(Tributylstannyl)pyridin
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
2-(Tri-n-butylstannyl)pyridine
011011-5G
011011-25G
FLUO011011-5GEA
119
EUR
FLUO011011-5G
FLUO011011-25G
2-(Tributylstannyl)pyridin
2-(Tributylstannyl)pyridin
Formel:
C₁₇H₃₁NSn Molecular Weight: 368,15 g/mol |
MDL:
MFCD00052052 CAS-Nummer: 17997-47-6 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...