S-Methylisothiouroniumsulfat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate
M-Clarity™ Qualitätsstufe = MQ100
M84445-100G
M84445-500G
SIALM84445-100GEA
32.4
EUR
SIALM84445-100G
SIALM84445-500G
S-Methylisothiouroniumsulfat, Sigma-Aldrich®
S-Methylisothiouronium sulphate
Formel:
C₄H₁₂N₄S₂·H₂SO₄ Molecular Weight: 278,38 g/mol |
MDL:
MFCD00013055 CAS-Nummer: 867-44-7 EINECS: 212-759-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...