tert-Butylisocyanid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: t-Butylisocyanide

M-Clarity™ Qualitätsstufe = MQ200

260630-1G 260630-5G
SIAL260630-1GEA 44.8 EUR
SIAL260630-1G SIAL260630-5G
tert-Butylisocyanid, Sigma-Aldrich®
tert-Butylisocyanid
Formel: C₅H₉N
Molecular Weight: 83,13 g/mol
MDL: MFCD00000002
CAS-Nummer: 7188-38-7
EINECS: 230-553-5
UN: 1992
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR