Cyclohexylisocyanid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

133302-1G 133302-5G
SIAL133302-1GEA 59.6 EUR
SIAL133302-1G SIAL133302-5G
Cyclohexylisocyanid, Sigma-Aldrich®
Cyclohexylisocyanid
Formel: C₇H₁₁N
MDL: MFCD00003839
CAS-Nummer: 931-53-3
EINECS: 213-238-7
UN: 2929
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR