Tetracyclin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T3258-5G T3258-100G T3258-25G
SIALT3258-5GEA 29.9 EUR
SIALT3258-5G SIALT3258-100G SIALT3258-25G
Tetracyclin, Sigma-Aldrich®
Tetracyclin
Formel: C₂₂H₂₄N₂O₈
Molecular Weight: 444.43 g/mol
MDL: MFCD00151232
CAS-Nummer: 60-54-8
EINECS: 200-481-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR