3,5-Difluor-4-methoxybenzylbromid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H26526.03
H26526.06
H26526.03EA
89.2
EUR
H26526.03
H26526.06
3,5-Difluor-4-methoxybenzylbromid ≥97%
3,5-Difluor-4-methoxybenzylbromid
Purity: 97%
CAS No.: 706786-42-7
Molecular Formula: C8H7BrF2O
Formula Weight: 237.04
Melting Point: 39-42°
UN No.: 3261
MDL No.: MFCD04115916
Formel:
C₈H₇BrF₂O Molecular Weight: 237.04 g/mol Schmelzpunkt: 39…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115916 CAS-Nummer: 706786-42-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...