3-Fluor-4-methoxybenzylbromid

Lieferant: FLUOROCHEM

Synonyme: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene

013839-1G 013839-5G 013839-25G
FLUO013839-1GEA 31.1 EUR
FLUO013839-1G FLUO013839-5G FLUO013839-25G
3-Fluor-4-methoxybenzylbromid
3-Fluor-4-methoxybenzylbromid
Formel: C₈H₈BrFO
MDL: MFCD00671768
CAS-Nummer: 331-61-3
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR