N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
M-Clarity™ Qualitätsstufe = MQ200
T7763-25G
T7763-5G
SIALT7763-25GEA
466
EUR
SIALT7763-25G
SIALT7763-5G
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14), Sigma-Aldrich®
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14)
Formel:
C₁₉H₄₁NO₃S Molecular Weight: 363.5988 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036910 CAS-Nummer: 14933-09-6 EINECS: 239-003-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...