3-Methylglutarsäureanhydrid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Methyldihydro-2H-pyran-2,6(3H)-dione
M47809-25G
M47809-5G
SIALM47809-25GEA
328
EUR
SIALM47809-25G
SIALM47809-5G
3-Methylglutarsäureanhydrid, Sigma-Aldrich®
3-Methylglutarsäureanhydrid
Formel:
C₆H₈O₃ Molecular Weight: 128,1259 g/mol |
MDL:
MFCD00006683 CAS-Nummer: 4166-53-4 EINECS: 224-020-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...