(+)-Dimethyl-D-malat zur Synthese
Lieferant: Merck
Synonyme: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate
8.18523.0001 8.18523.0005
8.18523.0001EA 0 EUR
8.18523.0001 8.18523.0005
(+)-Dimethyl-D-malat zur Synthese
(+)-Dimethyl-D-malat
Formel: C₆H₁₀O₅
Molecular Weight: 162,1406 g/mol
MDL: MFCD00066216
CAS-Nummer: 70681-41-3