(-)-Dimethyl-L-malat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-(-)-Dimethyl Malate, (S)-Dimethyl 2-hydroxysuccinate, Dimethyl (S)-(-)-malate
M-Clarity™ Qualitätsstufe = MQ200
374318-25G
374318-5G
SIAL374318-25GEA
185
EUR
SIAL374318-25G
SIAL374318-5G
(-)-Dimethyl-L-malat, Sigma-Aldrich®
(-)-Dimethyl-L-malat
Formel:
C₆H₁₀O₅ Molecular Weight: 162,1406 g/mol |
MDL:
MFCD00066215 CAS-Nummer: 617-55-0 EINECS: 432-310-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...