AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid), Millipore®
Lieferant: Merck Millipore Calbiochem
Achtung
Synonyme: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
101500-1 101500-100 101500-50 101500-500
101500-1EA 1010 EUR
101500-1 101500.100 101500-50 101500.500
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid), Millipore®
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Formel: C₈H₁₁ClFNO₂S
Molecular Weight: 239,69 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00132962
CAS-Nummer: 30827-99-7
UN: 3261
ADR: 8,II