Guanosin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

411130250 411131000 411135000
ACRO411130250EA 66.2 EUR
ACRO411130250 ACRO411131000 ACRO411135000
Guanosin 99%
Guanosin
Formel: C₁₀H₁₃N₅O₅
Molecular Weight: 283,24 g/mol
Schmelzpunkt: 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010182
CAS-Nummer: 118-00-3
EINECS: 204-227-8
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 ≥98.5 % (On dry substance)
Specific optical rotation -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR