Guanosin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
G6752-100G
G6752-25G
G6752-500G
SIALG6752-100GEA
342
EUR
SIALG6752-100G
SIALG6752-25G
SIALG6752-500G
Guanosin, Sigma-Aldrich®
Guanosin
Formel:
C₁₀H₁₃N₅O₅ Molecular Weight: 283,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010182 CAS-Nummer: 118-00-3 EINECS: 204-227-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...