Cytidin ≥99%
Lieferant: Thermo Fisher Scientific
A10261.09
A10261.18
A10261.30
A10261.09EA
57.7
EUR
A10261.09
A10261.18
A10261.30
Cytidin ≥99%
Cytidin
Purity: 99%
CAS No.: 65-46-3
Molecular Formula: C9H13N3O5
Formula Weight: 243.22
Melting Point: ca 215° dec.
Beilstein Reference: 89173
Merck Reference: 14,2786
Optical Rotation: +29° (c=9 in water)
MDL No.: MFCD00006545
Formel:
C₉H₁₃N₃O₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006545 CAS-Nummer: 65-46-3 EINECS: 200-610-9 Merck Index: 14,2786 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...