Cytidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

M-Clarity™ Qualitätsstufe = MQ200

C122106-10G C122106-1G
SIALC122106-10GEA 80 EUR
SIALC122106-10G SIALC122106-1G
Cytidin, Sigma-Aldrich®
Cytidin
Formel: C₉H₁₃N₃O₅
Molecular Weight: 243,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006545
CAS-Nummer: 65-46-3
EINECS: 200-610-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR