Cytidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
M-Clarity™ Qualitätsstufe = MQ100
C4654-1G
C4654-50G
C4654-5G
SIALC4654-1GEA
33.6
EUR
SIALC4654-1G
SIALC4654-50G
SIALC4654-5G
Cytidin, Sigma-Aldrich®
Cytidin
Formel:
C₉H₁₃N₃O₅ Molecular Weight: 243,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006545 CAS-Nummer: 65-46-3 EINECS: 200-610-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...