Uridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

U3750-100G U3750-25G
SIALU3750-100GEA 613 EUR
SIALU3750-100G SIALU3750-25G
Uridin, Sigma-Aldrich®
Uridin
Formel: C₉H₁₂N₂O₆
Molecular Weight: 244,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006526
CAS-Nummer: 58-96-8
EINECS: 200-407-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR