2'-Deoxyuridin 99+%
Lieferant: Thermo Scientific
|
Achtung
|
226610010
226612500
226610050
ACRO226610010EA
71
EUR
ACRO226610010
ACRO226612500
ACRO226610050
2'-Deoxyuridin 99+%
2'-Deoxyuridin
Formel:
C₉H₁₂N₂O₅ Molecular Weight: 228,20 g/mol Schmelzpunkt: 163…167 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006527 CAS-Nummer: 951-78-0 EINECS: 213-455-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
HPLC | ≥99.0 % |
Titration with NaOH | ≥99.0 % (On dry substance) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...