5-Bromuridin ≥98%

Lieferant: Thermo Scientific

A18507.01 A18507.03
A18507.01EA 26.3 EUR
A18507.01 A18507.03
5-Bromuridin ≥98%
5-Bromuridin

Purity: 98% CAS No.: 957-75-5 Molecular Formula: C9H11BrN2O6 Formula Weight: 323.10 Melting Point: 191-193° MDL No.: MFCD00006528

Formel: C₉H₁₁BrN₂O₆
Molecular Weight: 323,10 g/mol
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006528
CAS-Nummer: 957-75-5
EINECS: 213-486-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR