8-Bromguanosin

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

002895-25G 002895-100G 002895-5G
FLUO002895-25GEA 0 EUR
FLUO002895-25G FLUO002895-100G FLUO002895-5G
8-Bromguanosin
8-Bromguanosin
Formel: C₁₀H₁₂BrN₅O₅
Molecular Weight: 362.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037985
CAS-Nummer: 4016-63-1
EINECS: 223-677-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR