Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Lieferant: JENA BIOSCIENCES
Synonyme:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
NU-1013-100
NU-1013-1G
JENBNU-1013-100EA
0
EUR
JENBNU-1013-100
JENBNU-1013-1G
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Formel:
C₉H₁₂N₂Na₃O₁₅P₃ Molecular Weight: 550.09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00044310 CAS-Nummer: 19817-92-6 EINECS: 243-347-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...