Thiaminmonophosphatchlorid Dihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
T8637-5G
T8637-25G
SIALT8637-5GEA
61.8
EUR
SIALT8637-5G
SIALT8637-25G
Thiaminmonophosphatchlorid Dihydrat, Sigma-Aldrich®
Thiaminmonophosphatchlorid Dihydrat
Formel:
C₁₂H₁₈ClN₄O₄PS·2H₂O |
MDL:
MFCD00063473 CAS-Nummer: 273724-21-3 EINECS: 208-536-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...