Ammoniumchlorid 0.1 mol/l (0.1 N) aqueous solution

Lieferant: Bernd Kraft

Synonyme: Salmiak

20077.2000 20077.3000 20077.5300
KRAF20077.2000EA 69.6 EUR
KRAF20077.2000 KRAF20077.3000 KRAF20077.5300
Ammoniumchlorid 0.1 mol/l (0.1 N) aqueous solution
Ammoniumchlorid
Formel: NH₄Cl
Molecular Weight: 53.49 g/mol
Schmelzpunkt: 335
Lagertemperatur: Raumtemperatur
MDL: MFCD00011420
CAS-Nummer: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR