β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂)

Lieferant: Serva

30312.01 30312.02 30312.03
SERA30312.01EA 47.9 EUR
SERA30312.01 SERA30312.02 SERA30312.03
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂)
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na2)
Formel: C₂₁H₂₇N₇Na₂O₁₄P₂
MDL: MFCD00036200
CAS-Nummer: 606-68-8
EINECS: 210-123-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR