6,7-Dimethoxy-1H,3H-chinazolin-2,4-dion

Lieferant: FLUOROCHEM

Synonyme: 1,2,3,4-Tetrahydro-6,7-dimethoxyquinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-dione

024184-25G 024184-5G
FLUO024184-25GEA 288 EUR
FLUO024184-25G FLUO024184-5G
6,7-Dimethoxy-1H,3H-chinazolin-2,4-dion
6,7-Dimethoxy-1H,3H-chinazolin-2,4-dion
Formel: C₁₀H₁₀N₂O₄
Molecular Weight: 222,2 g/mol
MDL: MFCD00023889
CAS-Nummer: 28888-44-0
EINECS: 249-288-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR