9-Brom-1-nonanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Bromononan-1-ol

M-Clarity™ Qualitätsstufe = MQ100

448729-25G 448729-5G
SIAL448729-25GEA 140 EUR
SIAL448729-25G SIAL448729-5G
9-Brom-1-nonanol, Sigma-Aldrich®
9-Bromo-1-nonanol
Formel: C₉H₁₉BrO
Molecular Weight: 223.15 g/mol
MDL: MFCD00063348
CAS-Nummer: 55362-80-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR