1,1,2,2-Tetrachlor-1,2-dimethyldisilan
Lieferant: CHEMPUR SILANE
SILS15280.1
SILS15280.5
SILNSILS15280.1EA
0
EUR
SILNSILS15280.1
SILNSILS15280.5
1,1,2,2-Tetrachlor-1,2-dimethyldisilan
1,1,2,2-Tetrachlor-1,2-dimethyldisilan
Formel:
C₂H₆Cl₄Si₂ |
MDL:
MFCD00192469 CAS-Nummer: 4518-98-3 EINECS: 224-844-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...